missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Cullin 4A Recombinant Rabbit Monoclonal Antibody (23GB790)
GREENER_CHOICE

Catalog No. MA553730
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA553730 100 μL
MA553731 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MA553730 Supplier Invitrogen™ Supplier No. MA553730
Only null left

Rabbit Recombinant Monoclonal Antibody

Cullin 4A Recombinant Monoclonal Antibody for Flow, ICC/IF, Western Blot

CUL4A (Cullin 4A) is the ubiquitin ligase component of a multimeric complex involved in the degradation of DNA damage-response proteins.

Specifications

Antigen Cullin 4A
Applications Flow Cytometry, Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone 23GB790
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene CUL4A
Gene Accession No. Q13619
Gene Alias 2810470J21Rik; AW495282; CUL 4A; Cul4a; CUL-4A; Cul4a protein; cullin 4A; cullin-4A; MGC36573; MGC64071; OTTHUMP00000018753; RGD1563853
Gene Symbols CUL4A
Host Species Rabbit
Immunogen A synthetic peptide of human beta Cullin4A (700-759AA).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8451
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less