missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Cullin 3 Recombinant Rabbit Monoclonal Antibody (23GB1560)

Catalog No. MA552677
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA552677 20 μL
MA552676 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MA552677 Supplier Invitrogen™ Supplier No. MA552677

Rabbit Recombinant Monoclonal Antibody

Cullin 3 Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow

Cullin 3 is a member of the family of human cullin genes (CUL 1, 2, 3, 4a, 4b, and 5) homologous to the S. cerevisiae cdc 53 gene. CUL 1 forms a complex with human p19 skp1 and F box protein p45 skp2. Together with cdc 53 (Known as Cul A), as three subunits, they form a ubiquitin ligase-controlling proteolysis of G1 cell cycle regulatory proteins. In contrast to CUL 1, CUL 2 does not associate with p19 skp1 and p45 skp2. Instead, CUL 2 forms a complex with an inactive transcriptional elongation complex, SIII, formed by three subunits: Elongain C, Elongin B and VHL. The function of this association remains unknown.

Specifications

Antigen Cullin 3
Applications Flow Cytometry, Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone 23GB1560
Concentration 0.35 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene CUL3
Gene Accession No. B5DF89, Q13618, Q9JLV5
Gene Alias AI467304; AW146203; Cul3; CUL-3; cullin 3; cullin-3; KIAA0617; mKIAA0617; OTTHUMP00000203839; OTTHUMP00000203840; PHA2E
Gene Symbols CUL3
Host Species Rabbit
Immunogen A synthesized peptide derived from human Cullin 3 (700-768AA).
Purification Method Affinity chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26554, 301555, 8452
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less