missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cucurbitacin B 98.0+%, TCI America™
Supplier: TCI America C344225MG
Specifications
| Cucurbitacin B | |
| 179°C | |
| C32H46O8 | |
| 25 mg | |
| IXQKXEUSCPEQRD-DKRGWESNSA-N | |
| [(E,6R)-6-[(2S,8S,9R,10R,13R,14S,16R,17R)-2,16-dihydroxy-4,4,9,13,14-pentamethyl-3,11-dioxo-2,7,8,10,12,15,16,17-octahydro-1H-cyclopenta[a]phenanthren-17-yl]-6-hydroxy-2-methyl-5-oxohept-3-en-2-yl] acetate | |
| 5281316 | |
| 558.71 | |
| Crystalline Powder |
| 6199-67-3 | |
| White-Yellow | |
| MFCD07778083 | |
| 2811 | |
| CC(=O)OC(C)(C)C=CC(=O)C(C)(C1C(CC2(C1(CC(=O)C3(C2CC=C4C3CC(C(=O)C4(C)C)O)C)C)C)O)O | |
| 558.712 | |
| CHEBI:3941 | |
| ≥98.0% (HPLC) |
Chemical Identifiers
| 6199-67-3 | |
| 558.712 | |
| IXQKXEUSCPEQRD-DKRGWESNSA-N | |
| CHEBI:3941 | |
| CC(=O)OC(C)(C)C=CC(=O)C(C)(C1C(CC2(C1(CC(=O)C3(C2CC=C4C3CC(C(=O)C4(C)C)O)C)C)C)O)O |
| C32H46O8 | |
| MFCD07778083 | |
| 5281316 | |
| [(E,6R)-6-[(2S,8S,9R,10R,13R,14S,16R,17R)-2,16-dihydroxy-4,4,9,13,14-pentamethyl-3,11-dioxo-2,7,8,10,12,15,16,17-octahydro-1H-cyclopenta[a]phenanthren-17-yl]-6-hydroxy-2-methyl-5-oxohept-3-en-2-yl] acetate |
Safety and Handling
RTECSNumber : RC6190000
TSCA : No
Recommended Storage : Refrigerator