missing translation for 'onlineSavingsMsg'
Learn More

Sartorius Cubis III Precision Balance, Draft Shield R QP99

Catalog No. 14561500
Change view
Click to view available options
Capacity:
320 g
620 g
1.2 kg
2.2 kg
3.2 kg
5.2 kg
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity
14561500 620 g
14561461 320 g
14561432 1.2 kg
14561401 2.2 kg
14561441 3.2 kg
14561409 5.2 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 14561500 Supplier Sartorius Supplier No. CUB623S1S2RC

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Electronic precision balance with 7 in. color TFT touch display.

  • Deliver precise, reliable weighing results for virtually any regulated lab application, with capacities from 2.1 g to 70 kg and readabilities from 0.1 μg to 1 g.
  • A flexible range of weighing modules, draft shields, and software options lets customers tailor the balances to their workflow, compliance, and integration needs.
  • Upgradeable hardware and software protect your investment and keep your solution ready for future requirements.
  • Built for digital labs, Cubis III enables secure, plug-and-play integration with LIMS, ELN, and other lab systems via standards such as OPC UA, LADS, and SiLA.
  • This helps simplify onboarding of new balances, accelerates validation, and reduces integration costs, while providing a scalable foundation for automation.
  • As connectivity becomes essential to GxP operations, Cubis III delivers real business value by strengthening data integrity, improving audit readiness, accelerating batch release, reducing deviations, and lowering costs.
  • Fits seamlessly into connected labs for a more efficient, integrated, future-ready workflow.

Specifications

Capacity 620 g
Display Type Color TFT Touchscreen
Readability 1 mg
Includes Draft Shield R QP99
Type Precision Balance