missing translation for 'onlineSavingsMsg'
Learn More

Sartorius Cubis III High-capacity Balance, QP99

Catalog No. 14561553
Change view
Click to view available options
Capacity:
11.2 kg
20.2 kg
36.2 kg
50.2 kg
70.2 kg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity
14561553 36.2 kg
14561393 11.2 kg
14561506 20.2 kg
14561464 50.2 kg
14561448 70.2 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 14561553 Supplier Sartorius Supplier No. CUB36201S1S20C

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Precision electronic high-capacity balance with 7 in. color TFT touch display.

  • Deliver precise, reliable weighing results for virtually any regulated lab application, with capacities from 2.1 g to 70 kg and readabilities from 0.1 μg to 1 g.
  • A flexible range of weighing modules, draft shields, and software options lets customers tailor the balances to their workflow, compliance, and integration needs.
  • Upgradeable hardware and software protect your investment and keep your solution ready for future requirements.
  • Built for digital labs, Cubis III enables secure, plug-and-play integration with LIMS, ELN, and other lab systems via standards such as OPC UA, LADS, and SiLA.
  • This helps simplify onboarding of new balances, accelerates validation, and reduces integration costs, while providing a scalable foundation for automation.
  • As connectivity becomes essential to GxP operations, Cubis III delivers real business value by strengthening data integrity, improving audit readiness, accelerating batch release, reducing deviations, and lowering costs.
  • Fits seamlessly into connected labs for a more efficient, integrated, future-ready workflow.

Specifications

Capacity 36.2 kg
Display Type Color TFT Touchscreen
Readability 100 mg
Includes QP99
Type High-capacity Balance