missing translation for 'onlineSavingsMsg'
Learn More

Sartorius Cubis™ II Weighing Pan

Catalog No. 01675404
Change view
Click to view available options
Diameter (Metric):
50 mm
90 mm
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Diameter (Metric)
01675404 50 mm
01675403 90 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01675404 Supplier Sartorius Supplier No. YWP093

This Weighing Pan is an accessory for Cubis™ II.

  • Cubis™ II Accessory

Specifications

Autoclavable Not Autoclavable
Bottom Shape Concave
Color Silver
Disposable Reusable
Material Stainless Steel
Shape Round
Sterility Non-sterile
Type Weighing Pan
Product Line Cubis II
Diameter (English) 2 in.
Diameter (Metric) 50 mm
Show More Show Less