missing translation for 'onlineSavingsMsg'
Learn More

Sartorius Cubis™ II Precision (3-place) Balance, MCA User Interface

With completely configurable hardware, software, and connectivity, Cubis™ II offers a high-performance balance that will align with your unique demands and compliance requirements.

Specifications

Certifications/Compliance 21 CFR PART 11 17025 USP 41
Connection Type 2 USB A ports, 1 USB B port, 1USP C port, 1-9 pin RS 232 connection, 1 ethernet port
Display Type Color TFT Touchscreen
Leveling Automation motorized
Material Die-cast Aluminum, Plastic PBT, Optiwhite Float Glass and Stainless Steel, Aluminum Trim
View More Specs

Products 12
Catalog Number Mfr. No. Capacity Includes Model Price Quantity  
Catalog Number Mfr. No. Capacity Includes Model Price Quantity  
14561055
View Documents
Sartorius
MCA323S2S00E
320 g Balance, Accessories, Power supply, Manual, Factory test report Draft shield E: Small glass N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
14561053
View Documents
Sartorius
MCA623S2S00E
620 g Balance, Accessories, Power supply, Manual, Factory test report Draft shield E: Small glass N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
14561255
View Documents
Sartorius
MCA623SES00
620 g Balance, Accessories, Power supply, Manual, Factory test report, QP99 software Draft shield E: Small glass N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
14561052
View Documents
Sartorius
MCA1203S2S00E
1.2 kg Balance, Accessories, Power supply, Manual, Factory test report Draft shield E: Small glass N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
14561229
View Documents
Sartorius
MCA1203SES00
1.2 kg Balance, Accessories, Power supply, Manual, Factory test report, QP99 software Draft shield E: Small glass N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
14561050
View Documents
Sartorius
MCA2203S2S00E
2.2 kg Balance, Accessories, Power supply, Manual, Factory test report Draft shield E: Small glass N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
14561049
View Documents
Sartorius
MCA3203S2S00E
3.2 kg Balance, Accessories, Power supply, Manual, Factory test report Draft shield E: Small glass N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
14561244
View Documents
Sartorius
MCA3203SES00
3.2 kg Balance, Accessories, Power supply, Manual, Factory test report, QP99 software Draft shield E: Small glass N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
14561047
View Documents
Sartorius
MCA5203S2S00E
5.2 kg Balance, Accessories, Power supply, Manual, Factory test report Draft shield E: Small glass N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
14561054
View Documents
Sartorius
MCA623P2S00E
150 g, 300 g, 620 g Balance, Accessories, Power supply, Manual, Factory test report Draft shield E: Small glass N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
14561051
View Documents
Sartorius
MCA2203P2S00E
1.01 kg, 2.2 kg Balance, Accessories, Power supply, Manual, Factory test report Draft shield E: Small glass N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
14561048
View Documents
Sartorius
MCA5203P2S00E
1.2 kg, 2.4 kg, 5.2 kg Balance, Accessories, Power supply, Manual, Factory test report Draft shield E: Small glass N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

  • Monolithical weighing system
  • Integrated climate sensors
  • Individual sample holders
  • Error-free operation
  • Individual QApp workflows
  • Motorized auto-leveling
  • Full end-to-end data integrity
  • 21 CFR Part 11 compliance
  • Integrated audit-trail
  • State-of-the-art user management
  • Oustanding service support
  • Integrated status center
  • Integrated service functions
  • Preventive maintenance on accredited standards
  • For all models up to a maximum capacity of 8.2 kg
  • Large 7-inch color touch TFT display in 16:9 format with new user interface
  • Software Factory installed basic set of essential weighing applications (license free) and packages with special weighing applications and function extensions (license required)
  • Operation Activated by touch key, touch-free using IR sensor

    (draft shield M) or gesture sensor (optional), learning capability

Warranty

10-year warranty

Specifications

Specifications

21 CFR PART 11 17025 USP 41
Color TFT Touchscreen
Die-cast Aluminum, Plastic PBT, Optiwhite Float Glass and Stainless Steel, Aluminum Trim
Precision Balance
240 mm
120/240 V
Class I
2 USB A ports, 1 USB B port, 1USP C port, 1-9 pin RS 232 connection, 1 ethernet port
Automation motorized
0°C to +40°C
10 years
122/284/373 mm
1.0 A
Videos
SDS
Documents

Documents