missing translation for 'onlineSavingsMsg'
Learn More

Sartorius Cubis II MTS Balance

Catalog No. 01288034
Change view
Click to view available options
Capacity:
120 g
8200 g
Readability:
0.01
10 mg
Description:
MCA Display including touch, scroll, swipe functionality with factory-installed essential weighing applications and diverse QPapp Packages for optional software extensions
MCE Display with factory-installed essential weighing applications
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity Readability Description
01288034 8200 g 10 mg MCE Display with factory-installed essential weighing applications
01288035 8200 g 10 mg MCA Display including touch, scroll, swipe functionality with factory-installed essential weighing applications and diverse QPapp Packages for optional software extensions
01288036 120 g 0.01 MCA Display including touch, scroll, swipe functionality with factory-installed essential weighing applications and diverse QPapp Packages for optional software extensions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 01288034 Supplier Sartorius Supplier No. MCE8202S0MTS

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

The combinability of the display unit, weighing module, draft shield, software packages for various applications and functions and a comprehensive range of accessories allows the individual adaptation of the Cubis™ II balance to all weighing tasks.

  • Leading performance.
  • Monolithical weighing system, integrated climate sensors, individual sample holders
  • Error-free operation.
  • Individual QApp workflows, motorized auto-leveling
  • The modular weighing system Cubis™ II can be individually configured from different components.

Specifications

Connection Type D-sub connector, 9-pin
Description MCE Display with factory-installed essential weighing applications
Capacity 8200 g
Display Type Color Touchscreen
Readability 10 mg
Material Die-cast Aluminum, Plastic, Optiwhite Float Glass and Stainless Steel
Model Cubis II MTS Balance
Stabilization Time 1 second
Type Balance
Electrical Requirements 00 to 240 VAC (±10%)
Legal for Trade Class II
Show More Show Less