missing translation for 'onlineSavingsMsg'
Learn More

Sartorius Cubis™ II Analytical Balance, MCE User Interface

Cubis™ II Analytical Balance features fully customizable hardware, software, and connectivity

Supplier:  Sartorius MCE224S2S00I

 View more versions of this product

Catalog No. 14561145

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

  • Monolithical weighing system
  • Integrated climate sensors
  • Individual sample holders
  • Error-free operation
  • Individual QApp workflows
  • Motorized auto-leveling
  • Full end-to-end data integrity
  • Integrated audit-trail
  • State-of-the-art user management
  • Excellent service support
  • Integrated status center
  • Integrated service functions
  • Preventive maintenance on accredited standards
  • TFT touch screen operation with readable display and easy to operate user interface
  • Software Factory installed basic set of essential weighing applications
  • Operation Activated by touch key, touch-free using IR sensor (draft shield M), or gesture sensor (optional) learning capability
Specifications

Specifications

2 USB A ports, 1 USB B port, 1USP C port, 1 to 9 pin RS 232 connection, 1 ethernet port
220 g
0.1 mg
± 0.13 mg
MCE224S2S00I
1 sec.
10 years
425 mm
9.8 in.
250 mm
120/240 V
Class I
Draft shield I - Motorized with ionizer
Color TFT Touchscreen
Balance, Accessories, Power supply, Manual, Factory test report, Draft Shield I - Motorized with Ionizer
Die-cast Aluminum, Plastic PBT, Optiwhite Float Glass and Stainless Steel, Aluminum Trim
± 0.05 mg
Analytical Balance
Analytical
240 mm
373 mm
8.2, 10 kg
100/240 V ±10%
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions