missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ CTS™ Versene™ Solution

Catalog No. A4239101
Encompass
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A4239101 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A4239101 Supplier Gibco™ Supplier No. A4239101

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CTS Versene Solution is a cGMP-manufactured EDTA solution for use as a gentle non-enzymatic cell dissociation reagent.

CTS Versene Solution is a cGMP-manufactured EDTA solution for use as a gentle non-enzymatic cell dissociation reagent. It is formulated as 0.2 g EDTA(Na4) per liter of phosphate buffered saline (PBS) (0.48 mM). It reduces the burden in qualifying reagents when transitioning from the research bench to the clinic.

Maintain viability over multiple passages

PSCs passaged with this cGMP manufactured EDTA dissociation reagent maintain viability.

Designed with transition in mind

CTS Versene Solution has the same formulation as RUO Versene Solution but with additional characterization and quality control testing. CTS Versene Solution is manufactured at a site that uses methods and controls that conform with cGMP for medical devices, 21 CFR Part 820. Our FDA-registered manufacturing site is ISO 13485-certified. Full documentation traceability is available for CTS Versene Solution.

Specifications

Content And Storage Store at 2–8°C and protect from light

Cell Type Pluripotent Stem Cells (PSCs)
Classification Animal Origin-free
Concentration 1X
Form Liquid
Product Type Cell Culture Dissociation Reagent
Sterility Sterile-filtered
Format Bottle
For Use With (Application) Cell and Gene Therapy Research, Development, and Manufacturing
Manufacturing Quality ISO 13485, MDSAP, FDA-registered, 21 CFR 820
Quantity 100 mL
Shipping Condition Room Temperature
Show More Show Less
What is CTS?

The Gibco Cell Therapy Systems (CTS) portfolio of cell and gene therapy products are GMP manufactured, safety tested, and backed by regulatory documentation to support your transition from discovery through clinical and commercial manufacturing. Through our CTS solutions, we are committed to helping customers streamline therapeutic development, minimize risk, and ease the burden on their quality systems.

For Research Use or Manufacturing of Cell, Gene, or Tissue-Based Products. CAUTION: Not intended for direct administration into humans or animals.