missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ CTS™ L-Glutamine (200 mM) (100X)

Catalog No. A4000225303
Change view
Click to view available options
Quantity:
100 mL
200 mL
Format:
Bag - BioProcess Container (BPC)
Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Format
A4000225303 200 mL Bag - BioProcess Container (BPC)
A4000225301 100 mL Bottle
A4000225302 100 mL Bag - BioProcess Container (BPC)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. A4000225303 Supplier Gibco™ Supplier No. A4000225303

Gibco CTS L‐Glutamine is a ready-to-use 200 mM stock solution that acts as a cell culture supplement. L-glutamine is an amino acid that is required for cell culture.

Gibco Cell Therapy Systems (CTS) L‐Glutamine is a ready-to-use 200-mM stock solution that acts as a cell culture supplement. L-glutamine is an amino acid that is required for cell culture. It is involved in the formation of purine and pyrimidine nucleotides, amino sugars, glutathione, L-glutamate, and other amino acids, as well as in protein synthesis and glucose production. Unlike most other amino acids, L-glutamine is not stable in solution. The rate at which degradation proceeds is a function of time, temperature, and pH.

Closed systems compatible
CTS L-Glutamine is available in bottle and bioprocess container formats. The bioprocess container chamber is constructed from Aegis 5-14 film and includes sterile, weldable PVC/C-Flex hybrid tubing with a female Luer lock that helps provide versatile connection options for integrating into a closed system manufacturing process.

CTS quality
The Gibco CTS product line enables you to reduce your burden in qualifying reagents during your transition from research to clinical applications.

Products are manufactured at a site that uses methods and controls that conform to cGMP for medical devices, 21 CFR Part 820. Our FDA-registered manufacturing sites are ISO 13485- and ISO 9001–certified. Gibco CTS products follow USP <1043> ancillary materials for cell, gene, and tissue-engineered products within the responsibilities applicable to a supplier.

Gibco CTS products are of high quality and are supplied with harmonized documentation such as certificates of analysis and certificates of origin.

Specifications

Content And Storage -20°C to -5°C. Protect from light.
Concentration 200 mM (100X)
Culture Type Mammalian and Stem Cell Culture
Form Clear, colorless solution
Product Type Cell Culture Supplement
Sterility Sterile-filtered
Sterilization Method 0.2 μm Sterile-filtered
Format Bag - BioProcess Container (BPC)
For Use With (Application) Cell and Gene Therapy Research, Development, and Manufacturing
Manufacturing Quality ISO13485, MDSAP, FDA-registered, 21CFR820
Quantity 200 mL
Shipping Condition Dry Ice
Show More Show Less

For Research Use or Manufacturing of Cell, Gene, or Tissue-Based Products. CAUTION: Not intended for direct administration into humans or animals.