Learn More
Gibco™ CTS™ HiFi Cas9 Protein
Description
Gibco CTS HiFi Cas9 Protein is our GMP-manufactured high-fidelity CRISPR-Cas9 protein, engineered to deliver reduced off-target effects while maintaining high on-target activity. This protein is designed and manufactured in compliance with the principles of GMP and to meet requirements of the standards for Ancillary Materials for Cell, Gene, and Tissue-Based Products. It is provided in a high-concentration and large-scale format for process development and clinical research applications.
Features of CTS HiFi Cas9 Protein include:
• Exceptional off-target profile—over 70% reduction in off-target effects in primary immune cells, compared to CTS TrueCut Cas9 (wild type) at tested loci
• Maintain on-target activity—achieve comparable on-target activity to CTS TrueCut Cas9 (wild type) in primary immune cells at tested loci
• Stringent manufacturing process—manufactured to meet the standards for Ancillary Materials for Cell, Gene, and Tissue-Based Products including USP , Ph.Eur. 5.2.12, and ISO 20399 following the principles of 21 CFR Part 820 in an FDA-registered manufacturing site
• Extensive safety testing—includes sterility, endotoxin, mycoplasma, residual nucleic acid, and protein testing
• Traceability documentation—COA, COO, Drug Master files, and Regulatory Support files* are available
CTS HiFi Cas9 Protein is a Streptococcus pyogenes high-fidelity Cas9 protein, produced by recombinant DNA technology in E. coli for genome editing with CRISPR technology. Cas9 protein forms a very stable RNP complex with the guide RNA (gRNA) component of the CRISPR-Cas9 system. Incorporation of NLS aids delivery to the nucleus, increasing the editing efficiency and on-target cleavage.
If you would like to evaluate our HiFi Cas9 protein or conduct small-scale research experiments, we recommend using our research-grade TrueCut HiFi Cas9 Protein (Cat. No. A50576).
*Country-specific Regulatory Support File process will be initiated upon request and will require additional documentation time.
Specifications
Specifications
| Concentration | 10 mg/mL |
| Quantity | 2.5 mg |
| Product Type | Cas9 Protein |
| Content And Storage | Store at –20°C. |
| Shipping Condition | Dry Ice |
| Expression System | E. coli |
| Form | Liquid |
| Research Category | Genome Editing |
| Manufacturing Quality | ISO 13485 |
Frequently Asked Questions (FAQs)
The Gibco Cell Therapy Systems (CTS) portfolio of cell and gene therapy products are GMP manufactured, safety tested, and backed by regulatory documentation to support your transition from discovery through clinical and commercial manufacturing. Through our CTS solutions, we are committed to helping customers streamline therapeutic development, minimize risk, and ease the burden on their quality systems. Learn more here.
The Limited Use Label Licenses are available on the individual product pages. For additional questions, please contact outlicensing@thermofisher.com.
To request traceability documentation for Gibco CTS HiFi Cas9 Protein (Cat. No. A54223, A54224) and Gibco CTS TrueCut Cas9 Protein (Cat. No. A45220, A45221), please follow the instructions at the following link: https://www.thermofisher.com/us/en/home/clinical/cell-gene-therapy/cell-therapy/cell-therapy-systems/cell-therapy-systems-support.html
Gibco CTS TrueCut Cas9 Protein (Cat. No. A45220, A45221) is a Streptococcus pyogenes Cas9 wild-type protein, produced by recombinant DNA technology in E. coli and Gibco CTS HiFi Cas9 Protein (Cat. No. A54223, A54224) is a Streptococcus pyogenes high-fidelity CRISPR Cas9 protein, produced by recombinant DNA technology in E. coli. Both proteins are designed and manufactured in compliance with GMP.
Both Gibco CTS HiFi Cas9 Protein (Cat. No. A54223, A54224) and Invitrogen TrueCut HiFi Cas9 Protein (Cat. No. A50574, A50575) are Streptococcus pyogenes high-fidelity CRISPR Cas9 proteins, produced by recombinant DNA technology in E. coli. Both proteins have the same sequence with added nuclear localization signal. Gibco CTS HiFi Cas9 Protein is manufactured to meet the standards for Ancillary Materials for Cell, Gene, and Tissue-Based Products whereas Invitrogen TrueCut HiFI Cas9 Protein is a research-grade product.