missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ CTS™ B-27™ Supplement XenoFree (50X)

Catalog No. a5047501
Change view
Click to view available options
Quantity:
10 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A5047501 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A5047501 Supplier Gibco™ Supplier No. A5047501
Only null left

CTS B-27 Supplement XenoFree is based on the classic serum-free B-27 supplement and designed to help meet regulatory requirements for cell therapy manufacturing.

CTS B-27 Supplement XenoFree is a cGMP-manufactured, serum-free supplement designed to meet regulatory requirements for cell therapy manufacturing. This supplement supports the growth, proliferation, and differentiation of various cell types derived from pluripotent stem cells (PSCs). Example applications include induction of human neural stem cells (hNSCs), expansion and proliferation of hNSCs, differentiation of hNSCs, and maintenance of mature differentiated neurons in culture.

CTS quality

Gibco CTS products are manufactured at a site that uses methods and controls that conform with cGMP for medical devices, 21 CFR Part 820. Our FDA-registered manufacturing sites are ISO 13485– and ISO 9001–certified.

Accompanied by documentation to support your regulatory filing

  • Access to our Drug Master File (DMF) or Regulatory Support File (RSF)
  • Traceability documentation—Certificate of Origin, Certificate of Analysis, and Safety Data Sheet

Reduced risk from animal-origin materials and human-derived components

CTS B-27 Supplement XenoFree is formulated with human-derived components and AOF recombinant components. To help reduce viral clearance risks, the use of human-derived components has been minimized, and human-derived components are limited to materials that help meet regulatory standards for donor qualification, viral testing, and viral inactivation.

CTS B-27 Supplement XenoFree was specifically designed for use in stem cell research and cell therapy applications by making targeted modifications to the classic serum-free B-27 Supplement. CTS B-27 Supplement XenoFree supports the robust growth, expansion and differentiation of stem cells, making it a valuable addition to any cell therapy protocol where cGMP standards are required.

Specifications

Content And Storage Store at -5°C to -20°C and protect from light.
Classification Serum-free, Xeno-free
Concentration 50X
Culture Type Cells derived from Pluripotent Stem Cells (PSCs)
Form Liquid
Product Type Cell Culture Supplement
Sterility Sterile-filtered
For Use With (Application) Cell and Gene Therapy Research, Development, and Manufacturing
Manufacturing Quality ISO 13485, MDSAP, FDA-registered, 21 CFR 820
Quantity 10 mL
Shipping Condition Dry Ice
Show More Show Less

For Research Use or Manufacturing of Cell, Gene, or Tissue-Based Products. CAUTION: Not intended for direct administration into humans or animals.