missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ B-27™ Supplement, XenoFree, minus insulin

Catalog No. A3695201
Encompass
Change view
Click to view available options
Quantity:
10 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A3695201 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A3695201 Supplier Gibco™ Supplier No. A3695201

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

B-27™ Supplement, XenoFree, minus insulin

B-27 Supplement XenoFree Minus Insulin is based on the classic B-27 Supplement formulation and contains no animal-derived components at the primary component level. The absence of insulin makes this formulation ideal for the growth of hippocampal and other CNS neurons in applications where the presence of insulin causes interference or in the derivation of cardiomyocytes and mesodermal cells from stem cells.

Quality

B-27 Supplement XenoFree Minus Insulin is manufactured at a site that uses methods and controls that conform with cGMP for medical devices, 21 CFR Part 820. Our FDA-registered manufacturing sites are ISO 13485– and ISO 9001–certified.

Formulated xeno-free

The preparation contains no animal-derived components at the primary component level.

Specifications

Content And Storage Store at -5°C to -20°C and protect from light.

Description Without Insulin
Cell Type Neural Cells
Classification Serum-free
Concentration 50 X
Culture Type Mammalian Cell Culture
Form Liquid
Product Type Cell Culture Supplement
Serum Level Serum-Free
Sterility Sterile
pH 6 to 8
Product Line B-27
Quantity 10 mL
Shipping Condition Dry Ice
Source Xeno-Free
Tested For Endotoxin, Performance, Sterility, Mycoplasma
Show More Show Less
Do you offer B-27 supplements with and without insulin?

Yes, we do offer B-27 Supplement, minus insulin (Cat. Nos. A1895601 and A1895602). All our other B-27 supplements do contain insulin.

Can I thaw B-27 Supplements in a 37 degrees C water bath or incubator?

We do not recommend thawing at 37 degrees C. Best results are observed upon thawing at room temperature for approximately 2 hrs or overnight at 2-8 degrees C.

How do I thaw B-27 Supplements?

We recommend thawing it at 4 degrees C overnight or at room temperature for 2 hours. We don't recommend thawing in a water bath or incubator.

Can I re-freeze B-27 Supplements into smaller aliquots for future use?

Yes, we recommend that you aliquot when you first thaw it and thereafter subject the aliquot to no more than 2 additional freeze/thaw cycles.

For Research Use Only. Not for use in diagnostic procedures.