missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CTPS Recombinant Rabbit Monoclonal Antibody (23GB1685)
GREENER_CHOICE

Catalog No. MA553734
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA553734 100 μL
MA553735 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MA553734 Supplier Invitrogen™ Supplier No. MA553734
Only null left

Rabbit Recombinant Monoclonal Antibody

CTPS Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow

There are two genes encoding members of a new family of type II integral membrane proteins. Both are ubiquitously expressed, and tissue-specific alternative mRNA initiation and splicing generate at least two major isoforms of each protein, with the smaller isoforms being truncated at the N-terminus. These proteins are called nesprin-l and -2 for nuclear _envelope _spectrin repeat, as they are characterized by the presence of multiple, clustered spectrin repeats, bipartite nuclear localization sequences and a conserved C-terminal, single transmembrane domain. Transient transfection of EGFP-fusion expression constructs demonstrated their localization to the nuclear membrane with a novel C-terminal, TM-domain-containing sequence essential for perinuclear localization. Nesprin-l is developmentally regulated in both smooth and skeletal muscle and is relocalized from the nuclear envelope to the nucleus and cytoplasm during C2Cl2 myoblast differentiation. Nesprins may function as 'dystrophins of the nucleus' to maintain nuclear organization and structura integrity.

Specifications

Antigen CTPS
Applications Flow Cytometry, Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone 23GB1685
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene CTPS1
Gene Accession No. P17812, P70698
Gene Alias cb1040; CTP synthase; CTP synthase 1; CTP synthase 1a; CTP synthase a; CTP synthetase 1; CTPS; ctps1; ctps1a; ctpsa; cytidine 5-prime triphosphate synthetase; cytidine 5'-triphosphate synthase; cytidine 5'-triphosphate synthetase; IMD24; UTP--ammonia ligase 1; wu:fb49e03; wu:fe17b03
Gene Symbols Ctps, CTPS1
Host Species Rabbit
Immunogen A synthesized peptide derived from human CTP synthase/CTPS (250-300AA).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1503, 313560, 51797
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less