missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CTNNA3 Recombinant Rabbit Monoclonal Antibody (23GB5125)

Rabbit Recombinant Monoclonal Antibody
Supplier: Invitrogen™ MA552796
Description
CTNNA3 Recombinant Monoclonal Antibody for Flow, ICC/IF, Western Blot
Catenins are proteins that connect cadherin/beta-catenin cell adhesion complexes to the actin cytoskeleton. Although originally discovered in testis, alphaT-catenin is expressed in other tissues, the highest levels being observed in heart. Immunohistochemical analysis showed human alphaTcatenin localization at intercalated discs of cardiomyocytes and in peritubular myoid cells of testis. It is proven that alphaT-catenin can functionally restore cell-cell adhesion in colon cancer cells lacking alpha-catenins. This indicates that this protein is involved in the formation of specific cell-cell contacts in specific types of muscle cells. The encoded protein has about the same predicted size (100 kDa) as other alpha-catenins to which it shows an overall amino acid identity of 57%. In cells transfected with alphaT-catenin cDNA, interaction with alpha-catenin was demonstrated by coimmunoprecipitation. The 892_24D2S antibody against alphaT-catenin does not show any cross reaction with alphaE-catenin or alphaNcatenin in western blot analysis.Chemical Identifiers
| CTNNA3 | |
| -20°C | |
| Recombinant Monoclonal | |
| Liquid | |
| IgG | |
| Human, Mouse, Rat | |
| CTNNA3 | |
| 4930429L08Rik; 4933408A16; Alpha T-catenin; Alpha-3 catenin; alpha-catenin-like protein; alpha-T-catenin; ARVD13; Cadherin-associated protein; catenin (cadherin associated protein), alpha 3; catenin (cadherin-associated protein), alpha 3; catenin alpha 3; catenin alpha-3; Catna3; CTNNA3; RGD1559455; RGD1562230; Vr22 | |
| CTNNA3 | |
| Primary | |
| Affinity chromatography |
| 23GB5125 | |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q65CL1, Q9UI47 | |
| 216033, 29119, 361839 | |
| A synthesized peptide derived from human CTNNA3 (700-895AA). | |
| Antibody |
Specifications
| CTNNA3 | |
| Recombinant Monoclonal | |
| Unconjugated | |
| CTNNA3 | |
| 4930429L08Rik; 4933408A16; Alpha T-catenin; Alpha-3 catenin; alpha-catenin-like protein; alpha-T-catenin; ARVD13; Cadherin-associated protein; catenin (cadherin associated protein), alpha 3; catenin (cadherin-associated protein), alpha 3; catenin alpha 3; catenin alpha-3; Catna3; CTNNA3; RGD1559455; RGD1562230; Vr22 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 216033, 29119, 361839 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 23GB5125 | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q65CL1, Q9UI47 | |
| CTNNA3 | |
| A synthesized peptide derived from human CTNNA3 (700-895AA). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |