missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CTH Recombinant Rabbit Monoclonal Antibody (23GB3805)
GREENER_CHOICE

Catalog No. MA552634
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA552634 100 μL
MA552635 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MA552634 Supplier Invitrogen™ Supplier No. MA552634
Only null left

Rabbit Recombinant Monoclonal Antibody

CTH Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow

This gene encodes a cytoplasmic enzyme in the trans-sulfuration pathway that converts cystathione derived from methionine into cysteine. Glutathione synthesis in the liver is dependent upon the availability of cysteine. Mutations in this gene cause cystathioninuria. Alternative splicing of this gene results in two transcript variants encoding different isoforms.

Specifications

Antigen CTH
Applications Flow Cytometry, Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone 23GB3805
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene CTH
Gene Accession No. P32929
Gene Alias 0610010I13Rik; AI314617; BC019483; CSE; Cth; cth protein; CTL target antigen; Cys3; cystathionase; cystathionase (cystathionine gamma-lyase); cystathionase isoform 1; cystathionine gamma-lyase; Cystathionine gamma-lyase-like protein; cysteine desulfhydrase; cysteine-protein sulfhydrase; EGK_00877; fb58d08; gamma-cystathionase; H; homoserine deaminase; homoserine dehydratase; LOC102117863; MGC9471; PRB-RA; probasin-related antigen; QccE-22305; synthase; Unknown (protein for MGC:85785); wu:fb48e03; wu:fb58d08; zgc:66001; zgc:85785
Gene Symbols CTH
Host Species Rabbit
Immunogen A synthetic peptide of human Cystathionase (1-55AA).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1491
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less