missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ CTAB, Molecular biology grade, Calbiochem™,

Supplier:  MilliporeSigma™ 219374100GM

Catalog No. 219374100GM


Only null left

Description

Description

Specifications

Specifications

White
100 g
C19H42BrN
Cetrimide, Cetyltrimethylammonium Bromide
[Br-].CCCCCCCCCCCCCCCC[N+](C)(C)C
364.46
CHEBI:3567
Molecular Biology
57-09-0
Glass Bottle
MFCD00011772
LZZYPRNAOMGNLH-UHFFFAOYSA-M
hexadecyltrimethylazanium bromide
5974
≥98% (TLC)

Chemical Identifiers

57-09-0
364.46
LZZYPRNAOMGNLH-UHFFFAOYSA-M
5974
hexadecyltrimethylazanium bromide
C19H42BrN
MFCD00011772
Cetrimide, Cetyltrimethylammonium Bromide
CHEBI:3567
[Br-].CCCCCCCCCCCCCCCC[N+](C)(C)C
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions