missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CSN5 Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories Inc A300014AT
Description
The recommended shelf life for this product is 1 year from date of receipt. Application Note: For IHC, epitope retrieval with Tris-EDTA pH 9.0 is recommended for FFPE tissue sections. Based on 100% sequence identity, this antibody is predicted to
JAB-1 is a 38 kDa protein which was originally described as a coactivator for cJUN. It is present in a large protein complex called the "Signalosome", which has been proposed to play a signaling role in mammalian cells. This protein complex appears to be analogous to the COP9 complex in plants, which is important in light-mediated plant development. Although the specific function of JAB-1 in this complex is unclear, it has recently been implicated in targeting proteins for degradation by the proteasome.Specifications
| CSN5 | |
| Polyclonal | |
| TBS with 0.1% BSA and 0.09% sodium azide | |
| O35864, Q92905 | |
| Cops5 | |
| Between 175 and 225 | |
| 20 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
| Immunohistochemistry, Western Blot | |
| Unconjugated | |
| COPS5 | |
| CSN5, JAB1, MGC3149, MOV-34, SGN5 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 10987, 26754 | |
| 4° C | |
| Liquid |