Learn More
Invitrogen™ Rat CSF2 (GM-CSF) ELISA Kit
ELISA
Supplier: Invitrogen™ ERCSF2
Description
The Rat CSF2 (GM-CSF) ELISA quantitates Rt CSF2 in rat serum, plasma, or cell culture medium. The assay will exclusively recognize both natural and recombinant Rt CSF2. Principle of the method The Rat CSF2 solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.
The GM-CSF gene encodes for a cytokine that regulates the production, differentiation, and function of macrophages and granulocytes. The active form of the protein exists as a homodimer in the extracellular space. This gene is located in a cluster of related genes at the chromosome region 5q31, which has been associated with interstitial deletions in the 5q- syndrome and acute myelogenous leukemia. Other genes in the cluster include interleukins 4, 5, and 13. The gene is involved in promoting tissue inflammation. Elevated levels of cytokines, including the one produced by this gene, have been observed in SARS-CoV-2 infected patients with acute respiratory distress syndrome. Mice that lack this gene or its receptor have been shown to develop pulmonary alveolar proteinosis.Chemical Identifiers
| P48750 | |
| 10 pg/mL | |
| HRP | |
| Cytokines & Receptors | |
| This ELISA kit shows no cross-reactivity with the following cytokines tested: rat CINC-2, CINC-3, CNTF, Fractalkine, IL-1 alpha, IL-1 beta, IL-4, IL-6, IL-10, IFN-gamma, Leptin, Lix, MCP-1, MIP-3 alpha, beta-NGF, TIMP-1, TNF-alpha. | |
| ELISA, Protein Assays, Protein Biology, Cancer Biology, Immunology, Neuroscience, Neurobiology, Protein Detection, Protein Analysis | |
| 116630 | |
| <12% | |
| Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers | |
| RUO | |
| Plasma 50 μL, Serum 50 μL, Supernatant 100 μL | |
| Gm-csf, Gmcsf | |
| 1 hrs 20 mins | |
| Solid-phase sandwich ELISA |
| 12.35 to 3000 pg/mL | |
| 10 pg/mL to 3000 pg/mL | |
| ELISA | |
| Plasma, Serum, Supernatant | |
| 96 assays | |
| Colorimetric Microplate Reader | |
| Gm-csf,Gmcsf | |
| <10 | |
| HRP | |
| Immunology | |
| 2°C to 8°C | |
| GM-CSF | |
| 4 hrs 45 mins |
Specifications
| P48750 | |
| 10 pg/mL | |
| Solid-phase sandwich ELISA | |
| ELISA | |
| Plasma, Serum, Supernatant | |
| 96 assays | |
| ELISA, Protein Assays, Protein Biology, Cancer Biology, Immunology, Neuroscience, Neurobiology, Protein Detection, Protein Analysis | |
| 116630 | |
| <12% | |
| Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers | |
| 96 Tests | |
| Immunology | |
| 2°C to 8°C | |
| GM-CSF | |
| 4 hrs 45 mins |
| 12.35 to 3000 pg/mL | |
| 10 pg/mL to 3000 pg/mL | |
| HRP | |
| Cytokines & Receptors | |
| This ELISA kit shows no cross-reactivity with the following cytokines tested: rat CINC-2, CINC-3, CNTF, Fractalkine, IL-1 alpha, IL-1 beta, IL-4, IL-6, IL-10, IFN-gamma, Leptin, Lix, MCP-1, MIP-3 alpha, beta-NGF, TIMP-1, TNF-alpha. | |
| Rat | |
| Colorimetric Microplate Reader | |
| Gm-csf,Gmcsf | |
| <10 | |
| HRP | |
| RUO | |
| Plasma 50 μL, Serum 50 μL, Supernatant 100 μL | |
| Gm-csf, Gmcsf | |
| 1 hrs 20 mins |