missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Crystal Violet, pure, indicator

Crystal Violet, C25H30ClN3, CAS Number-548-62-9, 90-94-8, Basic Violet 3, C.I. 42555, Gentian Violet, 100g, 13, 756, CHEBI:41688, 407.99, Pure, Danger, GHS H Statement: Very toxic to aquatic life with long lasting effects. Harmful if swallowed. Causes serious eye damage. May cause cancer. | CAS: 548-62-9 | C25H30ClN3 | 407.986 g/mol

Supplier:  Thermo Scientific Chemicals 212121000

 View more versions of this product

Catalog No. AC212121000

Only null left

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Indicator
Specifications

Chemical Identifiers

215.0°C
C25H30ClN3
13, 756
Basic Violet 3, C.I. 42555, Gentian Violet
ZXJXZNDDNMQXFV-UHFFFAOYSA-M
[4-[bis[4-(dimethylamino)phenyl]methylidene]cyclohexa-2,5-dien-1-ylidene]-dimethylazanium;chloride
11057
407.99
Glass bottle
548-62-9 , 90-94-8
MFCD00011750
15, 4430
Solubility in water: 16g/L (25°C). Other solubilities: soluble in chloroform, 1 g/10 mL alcohol, 1 g/15 mL glycerin, practically insoluble in ether
CN(C)C1=CC=C(C=C1)C(=C2C=CC(=[N+](C)C)C=C2)C3=CC=C(C=C3)N(C)C.[Cl-]
407.986
CHEBI:41688
Pure
Crystal Violet, Indicator, Pure

Specifications

Crystal Violet
Indicator, Pure
C25H30ClN3
13, 756
Basic Violet 3, C.I. 42555, Gentian Violet
ZXJXZNDDNMQXFV-UHFFFAOYSA-M
[4-[bis[4-(dimethylamino)phenyl]methylidene]cyclohexa-2,5-dien-1-ylidene]-dimethylazanium;chloride
11057
407.99
Glass bottle
215.0°C
548-62-9 , 90-94-8
MFCD00011750
15, 4430
Solubility in water: 16g/L (25°C). Other solubilities: soluble in chloroform, 1 g/10 mL alcohol, 1 g/15 mL glycerin, practically insoluble in ether
CN(C)C1=CC=C(C=C1)C(=C2C=CC(=[N+](C)C)C=C2)C3=CC=C(C=C3)N(C)C.[Cl-]
407.986
CHEBI:41688
Pure
100 g
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Very toxic to aquatic life with long lasting effects.
Harmful if swallowed.
Causes serious eye damage.
May cause cancer.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Avoid release to the environment.
Obtain special instructions before use.
IF exposed or concerned: Get medical advice/attention.
Do not eat, drink or smoke when using this product.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only