missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Crystal Violet
Description
Crystal Violet is used as an acid-base indicator, alcohol denaturant, biological stain, textile dye, and an indicator for copper salts. It is a component of gram staining in microorganisms and cell lines.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Crystal Violet is used as an acid-base indicator, alcohol denaturant, biological stain, textile dye, and an indicator for copper salts. It is a component of gram staining in microorganisms and cell lines.
Solubility
Miscible with water and chloroform. Partially miscible with alcohol and glycerol.
Notes
Incompatible with acids, oxidizing agents, alkali metals, strong oxidizing agents, ammonia, acid chlorides, acid anhydrides, reducing agents and peroxides.
Crystal Violet is used as an acid-base indicator, alcohol denaturant, biological stain, textile dye, and an indicator for copper salts. It is a component of gram staining in microorganisms and cell lines.
Solubility
Miscible with water and chloroform. Partially miscible with alcohol and glycerol.
Notes
Incompatible with acids, oxidizing agents, alkali metals, strong oxidizing agents, ammonia, acid chlorides, acid anhydrides, reducing agents and peroxides.
Specifications
Specifications
| Melting Point | ∼215°C (decomposition) |
| UN Number | UN3077 |
| Beilstein | 4077708 |
| Merck Index | 14,4395 |
| Solubility Information | Miscible with water and chloroform. Partially miscible with alcohol and glycerol. |
| Quantity | 500 g |
| Formula Weight | 407.99 |
| Chemical Name or Material | Crystal Violet |
Safety and Handling
| Hazard Category | H302-H318-H351 |
| Hazard Statement | GHS H Statement H318-H351-H302 Causes serious eye damage. Suspected of causing cancer. Harmful if swallowed. |
| Precautionary Statement | P201-P202-P264b-P270-P281-P301+P312-P305+P351+P338-P308+P313-P310-P330-P501c |
| DOTInformation | Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S. |
| EINECSNumber | 208-953-6 |
| RTECSNumber | BO9000000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only