missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Crystal poncaeu 6R, 90%, MP Biomedicals™
Specifications
Specifications
| Assay Percent Range | 0.9 |
| Color | Red |
| Quantity | 1 g |
| Physical Form | Powder |
| Formula Weight | 502.4 |
| Grade | Not Available |
| Chemical Name or Material | Crystal poncaeu 6R |