missing translation for 'onlineSavingsMsg'
Learn More

Cryptocyanine 98.0+%, TCI America™

Catalog No. C0426200MG
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C0426200MG 200 mg
C04261G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C0426200MG Supplier TCI America Supplier No. C0426200MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 4727-50-8
Molecular Formula C25H25IN2
Molecular Weight (g/mol) 480.393
MDL Number MFCD00011970
InChI Key CEJANLKHJMMNQB-UHFFFAOYSA-M
Synonym cryptocyanin, cryptocyanine, kryptocyanin, kryptocyanine, cryptocyanine iodide, kryptocyanine iodide, 1,1'-diethylquino-4-carbocyanine iodide, cryptocyanine o.a. 2, 1,4'-carbocyanine iodide, 1,4'-quinocarbocyanine iodide
PubChem CID 16219292
ChEBI CHEBI:51502
IUPAC Name (4E)-1-ethyl-4-[(Z)-3-(1-ethylquinolin-1-ium-4-yl)prop-2-enylidene]quinoline;iodide
SMILES CCN1C=CC(=CC=CC2=CC=[N+](C3=CC=CC=C23)CC)C4=CC=CC=C41.[I-]

Specifications

Color Green-Yellow
Quantity 200 mg
UN Number 2811
Formula Weight 480.39
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Cryptocyanine
RTECSNumber VC3676880
TSCA Yes