accessibility menu, dialog, popup

UserName

ICH3NNCH3(E/Z)(E/Z)

CAS RN 4727-50-8

IUPAC Name: 1-ethyl-4-[3-(1-ethyl-1,4-dihydroquinolin-4-ylidene)prop-1-en-1-yl]quinolin-1-ium iodide
Synonyms: cryptocyanine cryptocyanin kryptocyanine iodide kryptocyanin cryptocyanine iodide kryptocyanine
Molecular Weight (g/mol): 480.39
Molecular Formula: C25H25IN2
InChi Key: CEJANLKHJMMNQB-UHFFFAOYSA-M
SMILES: [I-].CCN1C=CC(=CC=CC2=C3C=CC=CC3=[N+](CC)C=C2)C2=CC=CC=C12
Percent Purity 
  • (2)
  • (3)
Physical Form 
  • (2)
  • (3)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)