missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cryptocyanine 98.0+%, TCI America™
Specifications
Specifications
| Color | Green-Yellow |
| Quantity | 1 g |
| UN Number | 2811 |
| Formula Weight | 480.39 |
| Percent Purity | ≥98.0% (HPLC) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | Cryptocyanine |
Safety and Handling
| RTECSNumber | VC3676880 |
| TSCA | Yes |