missing translation for 'onlineSavingsMsg'
Learn More

Eppendorf™ CryoStorage Vials, 3D SafeCode

Provides a 3-level coding of the SafeCode system (datamatrix code, barcode, and clear writing) to enable safe sample identification. Up to 96 pre-labeled vials are pre-racked and pre-capped (0.5 mL to 4.0 mL) for storage applications.

Supplier:  Eppendorf™ 0030079442

 View more versions of this product

Catalog No. 05413987


Only null left

Description

Description

  • 3-level coding for safe sample identification
  • ECC 200 error correction function enables safe code reading with up to 30 % damaged codes
  • Temperature range: -196 °C (gas phase of LN2) to +121 °C
  • Manufactured from high-quality polypropylene in class VIII cleanroom for highest sample purity
  • Pre-capped and pre-racked for comfortable handling
  • Additional information on vials available via Eppendorf website for documentation purposes
  • Direct import into Eppendorf eLABInventory and eLABJournal software for convenient handling
Specifications

Chemical Identifiers

Polypropylene
Yes
Coded Cryobank Vial System
Yes
2D SafeCode

Specifications

Polypropylene
4.0 mL
Yes
2D SafeCode
Coded Cryobank Vial System
Non-sterile
48
Yes
480 (10 Racks of 48 Vials)
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions