missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Cryopreserved Hepatocyte Recovery Medium (CHRM™)

Catalog No. CM7000
Encompass
Change view
Click to view available options
Quantity:
50 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CM7000 50 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CM7000 Supplier Gibco™ Supplier No. CM7000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Cryopreserved Hepatocyte Recovery Medium (CHRM) is a specialized medium designed to support the efficient recovery of Gibco cryopreserved human hepatocytes following thawing.

Cryopreserved Hepatocyte Recovery Medium (CHRM™) is a specialized medium designed to support the efficient recovery of Gibco cryopreserved human hepatocytes following thawing. Formulated to promote high cell viability and function, CHRM™ helps minimize cellular stress and maintain hepatocyte performance.

This medium is optimized for a wide range of applications requiring physiologically relevant hepatocyte function, including drug metabolism and toxicity testing (ADME), gene therapy research, and disease modeling. By supporting consistent cell quality, CHRM™ enables reproducible experimental results across workflows.

Specifications

Content And Storage Store in refrigerator (2–8°C) and protect from light.
Cell Type Hepatocyte Cells
Classification FBS-supplemented
Form Liquid
Product Type Hepatocyte Supplement
Sterility Sterile-filtered
Sterilization Method Sterile-filtered
With Additives Phenol Red
Product Line Gibco
Quantity 50 mL
Remarks Proprietary formulation provided by AP Sciences
Shipping Condition Wet Ice
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.