missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Cryopreserved Hepatocyte Plating Medium (CHPM™)

Catalog No. CM9000
Encompass
Change view
Click to view available options
Quantity:
50 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CM9000 50 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CM9000 Supplier Gibco™ Supplier No. CM9000

Cryopreserved Hepatocyte Plating Medium (CHPM)

Cryopreserved Hepatocyte Plating Medium (CHPM®) used for plating Gibco cryopreserved human hepatocytes for optimal attachment and monolayer formation.

For research use only. Not intended for human or animal therapeutic or diagnostic use.

Related Link

Learn more about our Hepatic Cell Culture reagents

Specifications

Content And Storage Store at 4°C
Cell Type Hepatocyte Cells
Classification FBS-supplemented
Form Liquid
Product Type Hepatocyte Supplement
Sterility Sterile
With Additives Phenol Red
Product Line Gibco
Quantity 50 mL
Remarks Proprietary formulation provided by AP Sciences
Shipping Condition Wet Ice
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.