missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Greiner Bio-One Cryo.s Round Bottom 2 mL Polypropylene Tubes with External Thread Cap
Suitable for storage of cell cultures, tissue samples, microbiological samples, material of human or animal origin as well as antibodies, RNA, DNA and protein samples.
$760.29 - $803.54
Specifications
| Diameter (Metric) | 12.4 mm |
|---|---|
| Product Type | Cryogenic Storage Tube |
| Capacity (Metric) | 2 mL |
| Bottom Shape | Round (Starfoot) |
| Material | Polypropylene |
| Catalog Number | Mfr. No. | Closure Color | Style | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Closure Color | Style | Price | Quantity | ||||
|
07000257
|
Greiner Bio-One
126263 |
Natural | ID Writing Area |
Case of 500 Each for $796.68
|
|
||||
|
07000259
|
Greiner Bio-One
126280 |
Red | ID Writing Area |
Case of 500 Each for $781.54
|
|
||||
|
07000258
|
Greiner Bio-One
126261 |
Natural | No Writing Area |
Case of 500 Each for $760.29
|
|
||||
|
07000434
|
Greiner Bio-One
126277 |
Green | ID Writing Area |
Case of 500 Each for $803.54
|
|
||||
|
07000435
|
Greiner Bio-One
126278 |
Yellow | ID Writing Area |
Case of 500 Each for $796.68
|
|
||||
|
07000436
|
Greiner Bio-One
126279 |
Blue | ID Writing Area |
Case of 500 Each for $781.61
|
|
||||
Description
- With Star foot base
- Without ID writing area
- Sterile
- Non-pyrogenic and non-toxic
- Includes 150 white cap inserts
- Polyethylener case
- Dia. x Height: 12.4 x 47mm
Specifications
| 12.4 mm | |
| 2 mL | |
| Polypropylene | |
| 47 mm | |
| Sterile |
| Cryogenic Storage Tube | |
| Round (Starfoot) | |
| External | |
| Polyethylene case | |
| Threaded |