missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Greiner Bio-One Cryo.s Round Bottom 2 mL Polypropylene Tubes with External Thread Cap
Suitable for storage of cell cultures, tissue samples, microbiological samples, material of human or animal origin as well as antibodies, RNA, DNA and protein samples.
Supplier: Greiner Bio-One 126277
Description
- With Star foot base
- Without ID writing area
- Sterile
- Non-pyrogenic and non-toxic
- Includes 150 white cap inserts
- Polyethylener case
- Dia. x Height: 12.4 x 47mm
Service Plan
No
Certifications
CE
Specifications
| Cryogenic Storage Tube | |
| Green | |
| Round (Starfoot) | |
| External | |
| 47 mm | |
| ID Writing Area | |
| Threaded |
| 12.4 mm | |
| 2 mL | |
| Polypropylene | |
| Starfoot Base, ID writing area and 150 white cap inserts | |
| Polyethylene case | |
| Sterile |