missing translation for 'onlineSavingsMsg'
Learn More

Greiner Bio-One Cryo.s Conical Bottom 1 mL Polypropylene Tubes with Internal Thread Cap
GREENER_CHOICE

Made of high-grade polypropylene which guarantees high thermal resistance.

Supplier:  Greiner Bio-One 123277

 View more versions of this product

Catalog No. 07000250


Only null left

Description

Description

  • Internal thread has silicone gasket
  • With ID writing area and star foot base
  • Sterile
  • Non-pyrogenic and non-toxic
  • Includes 150 white cap inserts
  • Dia. x Height: 12.5 x 42mm
Specifications

Chemical Identifiers

Polypropylene
Sterile

Specifications

12.5 mm
With writing area
1 mL
Polypropylene
Starfoot Base, ID writing area and 150 white cap inserts
Sterile
Cryogenic Storage Tube
Green
Conical
Internal
48 mm
Threaded
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions