missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CRYM Monoclonal Antibody (OTI1G7)
GREENER_CHOICE

Catalog No. PIMA525192
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA525192 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA525192 Supplier Invitrogen™ Supplier No. MA525192
Only null left

Mouse Monoclonal Antibody

CRYM Monoclonal Antibody for Western Blot, ICC/IF, IHC (P), Flow

Crystallins are separated into two classes: taxon-specific and ubiquitous. The former class is also called phylogenetically-restricted crystallins. The latter class constitutes the major proteins of vertebrate eye lens and maintains the transparency and refractive index of the lens. This gene encodes a taxon-specific crystallin protein that binds NADPH and has sequence similarity to bacterial ornithine cyclodeaminases. The encoded protein does not perform a structural role in lens tissue, and instead it binds thyroid hormone for possible regulatory or developmental roles. Multiple alternatively spliced transcript variants have been found for this gene.

Specifications

Antigen CRYM
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Monoclonal
Clone OTI1G7
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene CRYM
Gene Accession No. Q14894, Q9QYU4
Gene Alias CDK108; Crym; crystallin mu; crystallin, mu; DFNA40; ketimine reductase mu-crystallin; mu-crystallin homolog; Mu-crystallin-like protein; NADP-regulated thyroid-hormone binding protein; NADP-regulated thyroid-hormone-binding protein; similar to Mu-crystallin homolog (NADP-regulated thyroid-hormone binding protein); THBP; thiomorpholine-carboxylate dehydrogenase
Gene Symbols CRYM
Host Species Mouse
Immunogen Full length human recombinant protein of CRYM produced in HEK293T cell.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 117024, 1428
Target Species Human, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2b
Show More Show Less