missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Crotonylated Lysine Monoclonal Antibody (K16287_3A9), Invitrogen™

Catalog No. MA555992
Change view
Click to view available options
Quantity:
50 μL
200 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA555992 50 μL
MA556372 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MA555992 Supplier Invitrogen™ Supplier No. MA555992
Only null left

Mouse Monoclonal Antibody

A Post translational modification to the amino acid lysine. It is being studied for links to diseased states.

Specifications

Antigen Crotonylated Lysine
Applications Immunoprecipitation, Western Blot
Classification Monoclonal
Clone K16287_3A9
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide
Gene Alias Kcr; Lysine
Host Species Mouse
Immunogen Crotonyl Lysine peptide.
Purification Method Protein A
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Chemical
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less