missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Crotonylated Lysine Antibody Cocktail, Invitrogen™

Catalog No. MA556374
Change view
Click to view available options
Quantity:
50 μL
200 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA556374 200 μL
MA555614 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MA556374 Supplier Invitrogen™ Supplier No. MA556374
Only null left

Mouse Cocktail Antibody

A Post translational modification to the amino acid lysine. It is being studied for links to diseased states.

Specifications

Antigen Crotonylated Lysine
Applications Western Blot
Classification Cocktail
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide
Gene Alias Kcr; Lysine
Host Species Mouse
Immunogen Crotonyl Lysine peptide.
Purification Method Affinity chromatography
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Chemical
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less