missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Crotonoyl Chloride (cis- and trans- mixture) 95.0+%, TCI America™
$102.52 - $594.63
Chemical Identifiers
| CAS | 10487-71-5 |
|---|---|
| Molecular Formula | C4H5ClO |
| Molecular Weight (g/mol) | 104.533 |
| MDL Number | MFCD00000734 |
| InChI Key | RJUIDDKTATZJFE-NSCUHMNNSA-N |
| PubChem CID | 643131 |
| IUPAC Name | (E)-but-2-enoyl chloride |
| SMILES | CC=CC(=O)Cl |
Chemical Identifiers
| 10487-71-5 | |
| 104.533 | |
| RJUIDDKTATZJFE-NSCUHMNNSA-N | |
| (E)-but-2-enoyl chloride |
| C4H5ClO | |
| MFCD00000734 | |
| 643131 | |
| CC=CC(=O)Cl |
Specifications
| 10487-71-5 | |
| 125°C | |
| MFCD00000734 | |
| 2920 | |
| CC=CC(=O)Cl | |
| 104.533 | |
| 104.53 | |
| Liquid |
| Yellow | |
| C4H5ClO | |
| 25 g | |
| RJUIDDKTATZJFE-NSCUHMNNSA-N | |
| (E)-but-2-enoyl chloride | |
| 643131 | |
| ≥95.0% (GC) | |
| Crotonoyl Chloride (cis- and trans- mixture) |
Safety and Handling
EINECSNumber : (2)-3377
TSCA : Yes