accessibility menu, dialog, popup

UserName

CAS RN 10487-71-5

IUPAC Name: (2E)-but-2-enoyl chloride
Synonyms: 2-butenoyl chloride
Molecular Weight (g/mol): 104.53
Molecular Formula: C4H5ClO
InChi Key: RJUIDDKTATZJFE-NSCUHMNNSA-N
SMILES: C\C=C\C(Cl)=O
Percent Purity 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results