missing translation for 'onlineSavingsMsg'
Learn More

Crotonoyl Chloride (cis- and trans- mixture) 95.0+%, TCI America™

Catalog No. C0424500G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C0424500G 500 g
C042425G 25 g
C0424100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. C0424500G Supplier TCI America Supplier No. C0424500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 10487-71-5
Molecular Formula C4H5ClO
Molecular Weight (g/mol) 104.533
MDL Number MFCD00000734
InChI Key RJUIDDKTATZJFE-NSCUHMNNSA-N
PubChem CID 643131
IUPAC Name (E)-but-2-enoyl chloride
SMILES CC=CC(=O)Cl

Specifications

Color Yellow
Boiling Point 125°C
Quantity 500 g
UN Number 2920
Formula Weight 104.53
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material Crotonoyl Chloride (cis- and trans- mixture)
EINECSNumber (2)-3377
TSCA Yes