missing translation for 'onlineSavingsMsg'
Learn More

Medisca Cromolyn Sodium USP

Catalog No. 14186127
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
14186127 100 g
14185111 25 g
14185623 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 14186127 Supplier Medisca Supplier No. 030105
Get 1 Extra Chemical When You Buy 4
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Cromolyn Sodium is a white, odorless, crystalline powder. It is hygroscopic. Alternative name(s): cromoglycate, cromoglycic acid, sodium cromoglicate

Cromolyn Sodium is a white, odorless, crystalline powder. It is hygroscopic.

Alternative name(s): cromoglycate, cromoglycic acid, sodium cromoglicate

Specifications

Chemical Name or Material Cromolyn Sodium USP
Packaging Jar, 300mL, White, HDPE, Round, 63-400, TE Pano Neck
CAS 15826-37-6
Type Drug Standard
Molecular Weight (g/mol) 512.3326
Quantity 100 g
Percent Purity 98 to 101 %
Grade USP