missing translation for 'onlineSavingsMsg'
Learn More

Crocin (Gardenia Fruits Extract), TCI America™

Catalog No. C152725G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C152725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C152725G Supplier TCI America Supplier No. C152725G
Only null left

Chemical Identifiers

CAS 42553-65-1
Molecular Formula C44H64O24
Molecular Weight (g/mol) 976.972
MDL Number MFCD00017495
InChI Key SEBIKDIMAPSUBY-AWBRESQISA-N
Synonym Crocetin Digentiobiosyl Ester
PubChem CID 133562570
IUPAC Name 1-O-[(2R,3S,4R,5R,6S)-3,4,5-trihydroxy-6-[[(2S,3S,4R,5R,6S)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxymethyl]oxan-2-yl] 16-O-[(3S,4R,5R,6S)-3,4,5-trihydroxy-6-[[(2S,3S,4R,5R,6S)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxymethyl]oxan-2-yl] (2E,4E,
SMILES CC(=CC=CC=C(C)C=CC=C(C)C(=O)OC1C(C(C(C(O1)COC2C(C(C(C(O2)CO)O)O)O)O)O)O)C=CC=C(C)C(=O)OC3C(C(C(C(O3)COC4C(C(C(C(O4)CO)O)O)O)O)O)O

Specifications

Quantity 25 g
Formula Weight 976.97
Physical Form Crystalline Powder
Chemical Name or Material Crocin (Gardenia Fruits Extract)
RTECSNumber HL7380000
TSCA No