Learn More
AlphaProTech Critical Cover™ CoolOne™ Mask with Magic Arch™
Fluid resistant face masks feature an integrated Magic Arch™ support strip to hold mask away from the nose and mouth to create a larger breathing chamber.
$401.54 - $604.51
Specifications
Product Type Medical/Procedural Face Mask
ASTM Level ASTM Level 1
Color Blue
Material Polypropylene
Size Standard
View More Specs
Products 2
Catalog Number Mfr. No. Fastening Type Model Price Quantity  
Catalog Number Mfr. No. Fastening Type Model Price Quantity  
19-169-4063
View Documents
AlphaProTech
BL6155
Earloops BL 6155 N/A
Only null left
19-130-4183
View Documents
AlphaProTech
BL6055
Earloops BL 6055 N/A
Only null left
Description
Description
  • Fluid resistant face masks are free of pigments and dyes
  • Patented Magic Arch™ support strip holds mask away from the nose and mouth to create a larger breathing chamber
  • Two styles to choose from:
    • Soft knit earloops
    • Anti-fog ear loops for use in high humidity environments or with protective eyewear
  • ASTM F2100 Compliant Level 1 mask.
Specifications
Specifications
Medical/Procedural Face Mask
Blue
Standard
Pleated
Single-use
0.1 μm
CoolOne™
ASTM Level 1
Polypropylene
Non-sterile
3-ply
≥95% (PFE), ≥95% (BFE)
Fluid
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'