missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AlphaProTech Critical Cover™ ComforTech™ Coveralls
Lightweight, durable microporous material delivers protection against particles, biologicals, and fluids in controlled environments. Available in 3 different styles.
Supplier: AlphaProTech CVJ40223
Description
Material rapidly dissipates vapor and heat on a microscopic level which helps to keep wearer cool and comfortable throughout the workday.
- Material has static dissipative properties
- Exhibits an ultra-low particle count and 99.9% filtration efficiency
- Fluid and particulate barrier
- Serged seams
- Latex-free
- White
Specifications
| Cleanroom Coveralls | |
| Microporous Composite Fabric | |
| Single-use | |
| Elastic | |
| Serged | |
| Elastic wrist and ankle | |
| 25 in. | |
| 63.5 cm | |
| For use in controlled environments and barrier to particles, biologicals, and fluids with 99.9% filtration efficiency | |
| 86.36 cm | |
| Class 10-100,000 (ISO 4-8) |
| Large | |
| White | |
| Elastic | |
| Zipper | |
| Non-sterile | |
| Yes | |
| 29.5 in. | |
| 74.93 cm | |
| 34 in. | |
| ComforTech™ |