missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CRIPT Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA561144
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA561144 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA561144 Supplier Invitrogen™ Supplier No. PA561144
Only null left

Rabbit Polyclonal Antibody

Immunogen sequence: CEKKLGTVIT PDTWKDGARN TTESGGRKLN ENKALTSKKA RFDPYGKNKF STCRICKSSV HQPGSHYCQG C Highest antigen sequence identity to the following orthologs: Mouse - 99%, Rat - 99%.

Involved in the cytoskeletal anchoring of DLG4 in excitatory synapses.

Specifications

Antigen CRIPT
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene CRIPT
Gene Accession No. Q9P021
Gene Alias 1200020A08Rik; AA589469; CRIPT; CXXC repeat containing interactor of PDZ3 domain; cysteine-rich interactor of PDZ three; Cysteine-rich interactor of PDZ3; cysteine-rich PDZ-binding protein; HSPC139; postsynaptic protein CRIPT; SSMDF
Gene Symbols CRIPT
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human CRIPT. Recombinant protein control fragment (Product #RP-96057).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9419
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less