missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CRIP3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA557568
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA557568 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA557568 Supplier Invitrogen™ Supplier No. PA557568

Rabbit Polyclonal Antibody

Immunogen sequence: CYGALFGPRG VNIGGVGSYL YNPPTPSPGC TTPLSPSSFS PPRPRTGLPQ GKKSPPHMKT FTGETSLCPG CGEPVYFAEK VMSLGRNWHR P Highest antigen sequence identity to the following orthologs: Mouse - 81%, Rat - 85%.

Specifications

Antigen CRIP3
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene CRIP3
Gene Accession No. Q6Q6R5
Gene Alias bA480N24.2; chromosome 6 LIM domain only protein; CRIP3; CRP3; CRP-3; cysteine rich protein 3; cysteine-rich protein 3; h6LIMo; thymus LIM protein TLP-A; TLP; TLP-A
Gene Symbols CRIP3
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human CRIP3. Recombinant protein control fragment (Product #RP-94884).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 401262
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less