missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Supelco™ Crimp seals with PTFE/Silicone Septa Silver Aluminum Seal

Catalog No. 111011562
Change view
Click to view available options
Color:
Red, Silver
Red, White, Silver
Silver
White, Silver
No. per Pack:
100
1000
Diameter (Metric) Inner:
11 mm
13 mm
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color No. per Pack Diameter (Metric) Inner
111011562 White, Silver 100 11 mm
111008938 White, Silver 1000 11 mm
111008973 Red, White, Silver 1000 11 mm
111011554 Red, Silver 100 11 mm
111011637 Silver 100 13 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 111011562 Supplier MilliporeSigma™ Supelco™ Supplier No. 27214U
Only null left

Crimp seals with silver aluminum seal

  • May be used to make specialized reaction vessel, by fusing them to top autosampler vials, in order to test the accuracy and reproducibility of the tetrabutylammonium fluoride and iodobenzene diacetateTBAF/PhI(OAc)2 method for detecting water in organic solvents.
  • It may also be used to seal various samples, thereby isolating them from atmospheric CO2 for a period of at least 1 month.

Specifications

Diameter (Metric) Inner 11 mm
Color White, Silver
Material PTFE, Aluminum (Seal)
Septum Silicone
No. per Pack 100
Thickness (Metric) 1.0 mm