missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Supelco™ Crimp seals with PTFE/Silicone Septa Silver Aluminum Seal

Crimp seals with silver aluminum seal

Supplier:  MilliporeSigma™ Supelco™ 27360U

 View more versions of this product

Catalog No. 111011554


Only null left

Description

Description

  • May be used to make specialized reaction vessel, by fusing them to top autosampler vials, in order to test the accuracy and reproducibility of the tetrabutylammonium fluoride and iodobenzene diacetateTBAF/PhI(OAc)2 method for detecting water in organic solvents.
  • It may also be used to seal various samples, thereby isolating them from atmospheric CO2 for a period of at least 1 month.
Specifications

Specifications

11 mm
PTFE, Aluminum (Seal)
100
Red, Silver
Silicone
1.0 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions