Learn More
MilliporeSigma™ Supelco™ Crimp Seals with PTFE/Silicone Septa
Used as an air-tight seal, and prevents contamination
$214.00
Specifications
Material Silicone, PTFE, Aluminum (Seal)
View More Specs
Products 1
Catalog Number Mfr. No. No. per Pack Price Quantity & Availability  
Catalog Number Mfr. No. No. per Pack Price Quantity & Availability  
111010534
View Documents Promotion Details
MilliporeSigma™ Supelco™
508489
100
Each for $214.00
Only null left
Description
Description
  • Used to seal the samples during the analysis of BTEX in BTEX contaminated waters, using static headspace extraction followed by gas chromatography with photoionization detection (HS-GC-PID).
Specifications
Specifications
Silicone, PTFE, Aluminum (Seal)