missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cresol Red 98.0+%, TCI America™
$55.92 - $169.63
Chemical Identifiers
| CAS | 1733-12-6 |
|---|---|
| Molecular Formula | C21H18O5S |
| Molecular Weight (g/mol) | 382.43 |
| MDL Number | MFCD00005878 |
| InChI Key | OBRMNDMBJQTZHV-UHFFFAOYSA-N |
| Synonym | o-Cresolsulfonphthalein |
| PubChem CID | 73013 |
| ChEBI | CHEBI:86218 |
| IUPAC Name | 3,3-bis(4-hydroxy-3-methylphenyl)-3H-2,1λâ¶-benzoxathiole-1,1-dione |
| SMILES | CC1=CC(=CC=C1O)C1(OS(=O)(=O)C2=CC=CC=C12)C1=CC=C(O)C(C)=C1 |
Chemical Identifiers
| 1733-12-6 | |
| 382.43 | |
| OBRMNDMBJQTZHV-UHFFFAOYSA-N | |
| 73013 | |
| 3,3-bis(4-hydroxy-3-methylphenyl)-3H-2,1λâ¶-benzoxathiole-1,1-dione |
| C21H18O5S | |
| MFCD00005878 | |
| o-Cresolsulfonphthalein | |
| CHEBI:86218 | |
| CC1=CC(=CC=C1O)C1(OS(=O)(=O)C2=CC=CC=C12)C1=CC=C(O)C(C)=C1 |
Specifications
| 1733-12-6 | |
| MFCD00005878 | |
| OBRMNDMBJQTZHV-UHFFFAOYSA-N | |
| 3,3-bis(4-hydroxy-3-methylphenyl)-3H-2,1λâ¶-benzoxathiole-1,1-dione | |
| 73013 | |
| 382.43 | |
| 1 g | |
| Cresol Red |
| C21H18O5S | |
| o-Cresolsulfonphthalein | |
| CC1=CC(=CC=C1O)C1(OS(=O)(=O)C2=CC=CC=C12)C1=CC=C(O)C(C)=C1 | |
| 382.43 | |
| CHEBI:86218 | |
| ≥98.0% (HPLC) | |
| Crystal/Powder |
Safety and Handling
TSCA : Yes