missing translation for 'onlineSavingsMsg'
Learn More

Cresol Red 98.0+%, TCI America™

Supplier:  TCI America C04061G

 View more versions of this product

Catalog No. C04061G

Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Cresol Red
C21H18O5S
o-Cresolsulfonphthalein
CC1=CC(=CC=C1O)C1(OS(=O)(=O)C2=CC=CC=C12)C1=CC=C(O)C(C)=C1
382.43
CHEBI:86218
≥98.0% (HPLC)
Crystal/Powder
1733-12-6
MFCD00005878
OBRMNDMBJQTZHV-UHFFFAOYSA-N
3,3-bis(4-hydroxy-3-methylphenyl)-3H-2,1λ⁶-benzoxathiole-1,1-dione
73013
382.43
1 g

Chemical Identifiers

1733-12-6
382.43
OBRMNDMBJQTZHV-UHFFFAOYSA-N
73013
3,3-bis(4-hydroxy-3-methylphenyl)-3H-2,1λ⁶-benzoxathiole-1,1-dione
C21H18O5S
MFCD00005878
o-Cresolsulfonphthalein
CHEBI:86218
CC1=CC(=CC=C1O)C1(OS(=O)(=O)C2=CC=CC=C12)C1=CC=C(O)C(C)=C1
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions