missing translation for 'onlineSavingsMsg'
Learn More

Cresol Red Sodium Salt, Reagent Grade, LabChem™

Supplier:  LabChem LC135107

Catalog No. LC135107


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Cresol Red Sodium Salt
62625-29-0
C21H17O5SNa
Passes Test
Reagent
Sulfur compounds; Carbon monoxide; Carbon dioxide
10 g
250°C
100
Soluble in water
404.42
Amber Glass
Brown
Powder

Chemical Identifiers

62625-29-0
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
Solution is not hazardous.

GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.

EINECSNumber : 263-654-8

Recommended Storage : Room Temperature

SDS
Documents

Documents

Promotions

Promotions